Capillary column ferrules for gas chromatography
85% vespel, 15% graphite ferrules, short, ferrule ID 0,4 mm, column ID 0,45, 0,53 mm
Katalognr. HEWL5062-3512
Anbieter: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalognr. HEWL5062-3512
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
Documents
HEWL5062-3514_TDS_EN
Datasheet
Technische Daten
- Bezeichnung:85% vespel, 15% graphite ferrules, short, ferrule ID 0,4 mm, column ID 0,45, 0,53 mm
- Seq:05