Capillary column ferrules for gas chromatography
Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,3 mm, column ID 0,1 mm
Katalognr. HEWL5062-3507
Anbieter: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalognr. HEWL5062-3507
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
Technische Daten
- Bezeichnung:Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,3 mm, column ID 0,1 mm
- Seq:10