AuftragsverfolgungKontakt
Electron capture detectors (ECD)
ECD adapter end cap, stainless steel
  HEWL19233-20755
Electron capture detectors (ECD)
  HEWL19233-20755
Electron capture detectors (ECD)
Electron capture detectors (ECD)
ECD adapter end cap, stainless steel
1 * 1 ST
The micro ECD has a detection zone volume 10 times smaller than any other ECD with replicable inlet liner to decreasing column contamination of the cell.
  • Replaceable liner
The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.
  • Bezeichnung:
    ECD adapter end cap, stainless steel
  • Seq:
    01