Electron capture detectors (ECD)
ECD adapter liner, fused silica
Katalognr. HEWL19233-20625
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
The micro ECD has a detection zone volume 10 times smaller than any other ECD with replicable inlet liner to decreasing column contamination of the cell.
- Replaceable liner
The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.
Technische Daten
- Bezeichnung:ECD adapter liner, fused silica
- Seq:02