Split/splitless inlet seals
Septum retainer nut
Katalognr. HEWL18740-60835
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
These inlet seals have been developed to ensure that you have a consistent and inert surface to properly seal the inlet and prevent sample degradation.
- Very reproducible smooth surface, eliminating traces of machining grooves that can be the source of minute leaks
- High quality, leak-free seal
Delivery information: An inlet washer is provided with each inlet seal.
Technische Daten
- Bezeichnung:Septum retainer nut
- Seq:09
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.