Split/splitless inlet seals
Septum retainer nut for headspace
Katalognr. HEWL18740-60830
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
These inlet seals have been developed to ensure that you have a consistent and inert surface to properly seal the inlet and prevent sample degradation.
- Very reproducible smooth surface, eliminating traces of machining grooves that can be the source of minute leaks
- High quality, leak-free seal
Lieferinformationen: An inlet washer is provided with each inlet seal.
Technische Daten
- Bezeichnung:Septum retainer nut for headspace
- Seq:08