Electron capture detectors (ECD)
ECD wipe test kit
Catalog # HEWL18713-60050
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
The micro ECD has a detection zone volume 10 times smaller than any other ECD with replicable inlet liner to decreasing column contamination of the cell.
- Replaceable liner
The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.
Specifications
- Bezeichnung:ECD wipe test kit
- Seq:04