Electron capture detectors (ECD)
GC electron capture detector standard in iso-octane, 0,5 ml
Katalognr. HEWL18713-60040
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
The micro ECD has a detection zone volume 10 times smaller than any other ECD with replicable inlet liner to decreasing column contamination of the cell.
- Replaceable liner
The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.
Technische Daten
- Bezeichnung:GC electron capture detector standard in iso-octane, 0,5 ml
- Seq:06