AuftragsverfolgungKontakt
Electron capture detectors (ECD) | Avantor
Electron capture detectors (ECD)
GC electron capture detector standard in iso-octane, 0,5 ml
Katalognr. HEWL18713-60040
Electron capture detectors (ECD)
Katalognr. HEWL18713-60040
Electron capture detectors (ECD)
Electron capture detectors (ECD)
GC electron capture detector standard in iso-octane, 0,5 ml
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
The micro ECD has a detection zone volume 10 times smaller than any other ECD with replicable inlet liner to decreasing column contamination of the cell.
  • Replaceable liner
The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.
Technische Daten
  • Bezeichnung:
    GC electron capture detector standard in iso-octane, 0,5 ml
  • Seq:
    06