Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
Ferrule, VG-1 vespel, 15% graphite, 1/8" - 1/16"
  HEWL0100-1344
Capillary column ferrules for gas chromatography
  HEWL0100-1344
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
Ferrule, VG-1 vespel, 15% graphite, 1/8" - 1/16"
1 * 10 ST
Documents
  • Bezeichnung:
    Ferrule, VG-1 vespel, 15% graphite, 1/8" - 1/16"
  • Seq:
    53
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.