Jars with base, knobbed lid
80 mm, 120 mm
Katalognr. 216-4297
Anbieter: GLASWARENFABRIK KARL HECHT
Direkt zu
Product Details & Documents
Made of glass, with glass or stainless steel lid.
- Good chemical resistance
- Sturdy
- Easy to clean
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Technische Daten
- Ext.Ø:80 mm
- Höhe:120 mm