Jars with base, knobbed lid
100 mm, 200 mm
Catalog # HECH44410072
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Made of glass, with glass or stainless steel lid.
- Good chemical resistance
- Sturdy
- Easy to clean
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Specifications
- Ext.Ø:100 mm
- Höhe:200 mm