Jars with base, knobbed lid
100 mm, 150 mm
Katalognr. 216-4294
Anbieter: GLASWARENFABRIK KARL HECHT
Direkt zu
Product Details & Documents
Made of glass, with glass or stainless steel lid.
- Good chemical resistance
- Sturdy
- Easy to clean
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Technische Daten
- Ext.Ø:100 mm
- Höhe:150 mm