Retort rods, unthreaded
13 mm, 1500 mm
241-2117
Stainless steel.
- Brilliantly polished stainless steel with good surface hardness that will not score from clamp binding screws
- Hollow, lightweight rods (Ø 13 mm) with continuously welded walls (1,6 mm thick)
- Good chemical resistance
- Red caps
- Ø:13 mm
- Länge:1500 mm
- Seq:07



