Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Preparation knives

Preparation knives

Preparation knife, very delicate, 23 mm, 125 mm
Katalognr. 233-0088
Anbieter:   HAMMACHER KARL
Preparation knives
Katalognr. 233-0088
Anbieter:  HAMMACHER KARL
Preparation knives
Preparation knives

Preparation knife, very delicate, 23 mm, 125 mm

Preis
SFr. 109.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

WIRONIT steel.

Technische Daten
  • Typ:
    Preparation knife, very delicate
  • Seq:
    02
  • Gesamtlänge:
    125 mm
  • Klingen:
    23 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.