Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Schläuche, PTFE

Gauge 9, 3,00 mm, 3,76 mm
Catalog # HAMI20909
Supplier:   HAMILTON BONADUZ
Schläuche, PTFE
Catalog # HAMI20909
Supplier:  HAMILTON BONADUZ
Schläuche, PTFE
Schläuche, PTFE

Gauge 9, 3,00 mm, 3,76 mm

Price
SFr. 43.00
Log in for availability
1 * 1 ST
Product Details & Documents

Luer hub tubing assemblies.

Ideal for quickly connecting and disconnecting components with male luer locks.

Specifications
  • Schlauch Außen-Ø:
    3,76 mm
  • Schlauch Innen-Ø:
    3,00 mm
  • Bezeichnung:
    Gauge 9
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.