Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Schläuche, PTFE

Schläuche, PTFE

Gauge 7, 3,76 mm, 4,52 mm
Codice catalogo HAMI20907
Nome del fornitore:   HAMILTON BONADUZ
Schläuche, PTFE
Codice catalogo HAMI20907
Nome del fornitore:  HAMILTON BONADUZ
Schläuche, PTFE
Schläuche, PTFE

Gauge 7, 3,76 mm, 4,52 mm

Price
44.20 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Luer hub tubing assemblies.

Ideal for quickly connecting and disconnecting components with male luer locks.

Specifiche
  • Schlauch Außen-Ø:
    4,52 mm
  • Schlauch Innen-Ø:
    3,76 mm
  • Bezeichnung:
    Gauge 7
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.