AuftragsverfolgungKontakt
Test kits, digital titration method
Test kits, digital titration method
Hardness (total+calcium+mg) reagent set, low range, 10 - 160 mg/l, 100 Tests
Katalognr. HACH2448600
Anbieter:   Hach
Test kits, digital titration method
Katalognr. HACH2448600
Anbieter:  Hach
CAS Nummer:  
Test kits, digital titration method
Test kits, digital titration method
Hardness (total+calcium+mg) reagent set, low range, 10 - 160 mg/l, 100 Tests
CAS Nummer:  
Preis
300.00 SFr.
Einloggen für Verfügbarkeit
1 * 100 Tests
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Technische Daten
Technische Daten
  • VE:
    100 Tests
  • Bezeichnung:
    Hardness (total+calcium+mg) reagent set, low range
  • Messbereich:
    10 - 160 mg/l