Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Immersion oils

Immersion oils

Flasche 100 ml Immersionsöl*, Bottle, 100 mL
Katalognr. 630-0279
Anbieter:   ZEISS
Immersion oils
Katalognr. 630-0279
Anbieter:  ZEISS
CAS Nummer:  
Immersion oils
Immersion oils

Flasche 100 ml Immersionsöl*, Bottle, 100 mL

CAS Nummer:  
Preis
SFr. 148.20
Einloggen für Verfügbarkeit
1 * 100 mL
Product Details & Documents

Increases the resolving power of a microscope. Zeiss Immersol™ is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.

  • Bubble-free immersion oil
  • Suitable for oil immersion objectives from Carl Zeiss

Certifications: ISO 8036-1/2

Technische Daten

Technische Daten

  • Packung:
    Bottle
  • VE:
    100 mL
  • Bezeichnung:
    Flasche 100 ml Immersionsöl*

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.