Kinetex Bulk Packing
Phenyl-Hexyl, 5 µm, 100 Å
Catalog # PHEC04A-4603
New Product
Supplier: PHENOMENEX
Jump to:
Product Details & Documents
Specifications
Specifications
- Partikelgröße:5 µm
- Porengröße:100 Å
- Bezeichnung:Phenyl-Hexyl