Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Kinetex Bulk Packing
Kinetex Bulk Packing
Phenyl-Hexyl, 5 µm, 100 Å
Catalog # PHEC04A-4603
New Product
Supplier:   PHENOMENEX
Kinetex Bulk Packing
Catalog # PHEC04A-4603
Supplier:  PHENOMENEX
New Product
CAS Number:  
Kinetex Bulk Packing
Kinetex Bulk Packing
Phenyl-Hexyl, 5 µm, 100 Å
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
Specifications
  • Partikelgröße:
    5 µm
  • Porengröße:
    100 Å
  • Bezeichnung:
    Phenyl-Hexyl
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.