Kinetex Bulk Packing
C8, 1,7 µm, 100 Å
Katalognr. PHEC04A-4499
New Product
Anbieter: PHENOMENEX
Direkt zu
Product Details & Documents
Technische Daten
Technische Daten
- Partikelgröße:1,7 µm
- Porengröße:100 Å
- Bezeichnung:C8