Kinetex Bulk Packing
XB-C18, 1,7 µm, 100 Å
Catalog # PHEC04A-4498
New Product
Supplier: PHENOMENEX
Jump to:
Product Details & Documents
Specifications
Specifications
- Partikelgröße:1,7 µm
- Porengröße:100 Å
- Bezeichnung:XB-C18
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.