Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Kinetex Bulk Packing
Kinetex Bulk Packing
C18, 1,7 µm, 100 Å
Catalog # PHEC04A-4475
New Product
Supplier:   PHENOMENEX
Kinetex Bulk Packing
Catalog # PHEC04A-4475
Supplier:  PHENOMENEX
New Product
CAS Number:  
Kinetex Bulk Packing
Kinetex Bulk Packing
C18, 1,7 µm, 100 Å
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
Specifications
  • Partikelgröße:
    1,7 µm
  • Porengröße:
    100 Å
  • Bezeichnung:
    C18
Specifications