Vitamin E Acetate 99%
Bottle
Katalognr. APOSOR1015424-25KG
New Product
Anbieter: Apollo Scientific
Vitamin E Acetate 99%
Katalognr. APOSOR1015424-25KG
Anbieter: Apollo Scientific
New Product
CAS Nummer: 7695-91-2
Direkt zu
Product Details & Documents
Usage: Organic building block/ reagent.
- Store at 2 to 8 °C
- Non-hazardous
Synonyme: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate
- CAS Nummer:7695-91-2
- Formel:C₃₁H₅₂O₃
- MDL-Nummer:MFCD00072042
- Molekulargewicht:472,74 g/mol
- Lagertemperatur:Kühlschrank
- EINECS:231-710-0
Technische Daten
Technische Daten
- Packung:Bottle



