Methyl Tridecanoate 98%
Bottle
APOSOR10138852.5KG
New Product
Usage: Organic building block/ reagent.
: Tridecylic acid methyl ester, Tridecanoic acid methyl ester
- :1731-88-0
- :C₁₄H₂₈O₂
- :MFCD00008977
- :228,37 g/mol
- Packung:Bottle
Usage: Organic building block/ reagent.