Fluorescein 95%
Bottle
Codice catalogo APOSOR1013317-25KG
New Product
Nome del fornitore: Apollo Scientific
Fluorescein 95%
Codice catalogo APOSOR1013317-25KG
Nome del fornitore: Apollo Scientific
New Product
Numero CAS: 2321-07-5
Vai a
Product Details & Documents
Usage: Organic building block/reagent
Sinonimi: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
- Numero CAS:2321-07-5
- Formula:C₂₀H₁₂O₅
- Numero MDL:MFCD00005050
- Molecular Weight:332,31 g/mol
Specifiche
Specifiche
- Packung:Bottle
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



