Acetophenone 99%
Bottle
Catalog # APOSOR1012200-2.5L
New Product
Supplier: Apollo Scientific
Acetophenone 99%
Catalog # APOSOR1012200-2.5L
Supplier: Apollo Scientific
New Product
CAS Number: 98-86-2
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: Methyl phenyl ketone, Phenyl methyl ketone
- CAS Number:98-86-2
- Formula:C₈H₈O
- MDL Number:MFCD00008724
- Molecular Weight:120,15 g/mol
Specifications
Specifications
- Packung:Bottle