Methylnonadecanoate 98%
Bottle
Katalognr. APOSOR1013903-1G
New Product
Anbieter: Apollo Scientific
Methylnonadecanoate 98%
Katalognr. APOSOR1013903-1G
Anbieter: Apollo Scientific
New Product
CAS Nummer: 1731-94-8
Direkt zu
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyme: Nonadecanoic acid methyll ester, Methyl Nonadecanoate
- CAS Nummer:1731-94-8
- Formel:C₂₀H₄₀O₂
- MDL-Nummer:MFCD00009011
- Molekulargewicht:312,53 g/mol
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



