Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
4,4-Difluorocyclohexan-1-amine 97%
4,4-Difluorocyclohexan-1-amine 97%
Bottle
  APOSPC901499-25G
New Product
4,4-Difluorocyclohexan-1-amine 97%
  APOSPC901499-25G
New Product
 :  458566-84-2
4,4-Difluorocyclohexan-1-amine 97%
Bottle
 :  458566-84-2
SFr. 1’300.00
1 * 25 g
 :  4,4-Difluorocyclohexanamine
  •  :458566-84-2
  •  :C₆H₁₁F₂N
  •  :MFCD06409984
  •  :135.155 g/mol
  •  :Raumtemperatur
  • Packung:
    Bottle
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.