MOPS 99% für die Herstellung einer Pufferlösung
Bottle
Katalognr. APOSBIA1076-2.5KG
New Product
Anbieter: Apollo Scientific
MOPS 99% für die Herstellung einer Pufferlösung
Katalognr. APOSBIA1076-2.5KG
Anbieter: Apollo Scientific
New Product
CAS Nummer: 1132-61-2
Direkt zu
Product Details & Documents
Usage: Biochemical/life science applications.
- Store at 2 to 8 °C
Synonyme: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
- CAS Nummer:1132-61-2
- Formel:C₇H₁₅NO₄S
- MDL-Nummer:MFCD00006183
- Molekulargewicht:209,266 g/mol
- Lagertemperatur:Kühlschrank
- EINECS:214-478-5
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



