Leptomycin B
Bottle
Katalognr. APOSBIL2101-100MG
New Product
Anbieter: Apollo Scientific
Leptomycin B
Katalognr. APOSBIL2101-100MG
Anbieter: Apollo Scientific
New Product
CAS Nummer: 87081-35-4
Direkt zu
Product Details & Documents
Potent inhibitor of nuclear export.
Supplied as a 1mM solution in abs. ethanol
Synonyme: (2E,5S,6R,7S,9R,10E,12E,15R,16Z,18E)-17-Ethyl-6-hydroxy-3,5,7,9,11,15-hexamethyl-19-((2S,3S)-3-methyl-6-oxo-3,6-dihydro-2H-pyran-2-yl)-8-oxononadeca-2,10,12,16,18-pentaenoic acid
- CAS Nummer:87081-35-4
- Formel:C₃₃H₄₈O₆
- MDL-Nummer:MFCD06795848
- Molekulargewicht:540.73 g/mol
- Blitz Pt:11,1 °C
- Lagertemperatur:Gefrierschrank
- UN:1170
- ADR:3,II
Technische Daten
Technische Daten
- Packung:Bottle



