Actinomycin D ≥99%
Bottle
Catalog # APOSBIA0175-250MG
New Product
Supplier: Apollo Scientific
Actinomycin D ≥99%
Catalog # APOSBIA0175-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 50-76-0
Jump to:
Product Details & Documents
Antineoplastic antibiotic
- Purity: ≥99%
- CAS Number:50-76-0
- Formula:C₆₂H₈₆N₁₂O₁₆
- MDL Number:MFCD00005033
- Molecular Weight:1255.5 g/mol
- Melting Pt:251…253 °C
- Storage Temperature:Gefrierschrank
- EINECS:200-063-6
- UN:3462
- ADR:6.1,II
Specifications
Specifications
- Packung:Bottle



