Picolinamid 95%
Bottle
Katalognr. APOSOR937776-500G
New Product
Anbieter: Apollo Scientific
Picolinamid 95%
Katalognr. APOSOR937776-500G
Anbieter: Apollo Scientific
New Product
CAS Nummer: 1452-77-3
Direkt zu
Product Details & Documents
Synonyme: 2-Pyridinecarboxamide, Pyridine-2-carboxamide
- CAS Nummer:1452-77-3
- Formel:C₆H₆N₂O
- MDL-Nummer:MFCD00023483
- Molekulargewicht:122.13 g/mol
- Lagertemperatur:Raumtemperatur
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



