AuftragsverfolgungKontakt
Acetophenone ≥98%
Bottle
  APOSBUP18875-1G
New Product
Acetophenone ≥98%
  APOSBUP18875-1G
New Product
 :  98-86-2
Acetophenone ≥98%
Bottle
 :  98-86-2
86.60 SFr.
1 * 1 g
Usage: Biochemical/ life science applications.
 :  Methyl phenyl ketone, Phenyl methyl ketone
  •  :98-86-2
  •  :C₈H₈O
  •  :120,15 g/mol
  • Packung:
    Bottle