Acetophenone ≥98%
Bottle
Katalognr. APOSBUP18875-500MG
New Product
Anbieter: Apollo Scientific
Acetophenone ≥98%
Katalognr. APOSBUP18875-500MG
Anbieter: Apollo Scientific
New Product
CAS Nummer: 98-86-2
Direkt zu
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyme: Methyl phenyl ketone, Phenyl methyl ketone
- CAS Nummer:98-86-2
- Formel:C₈H₈O
- Molekulargewicht:120,15 g/mol
Technische Daten
Technische Daten
- Packung:Bottle



