Hygromycin B ≥98%
Bottle
APOSBUP17695-5G
New Product
Usage: Biochemical/life science applications
: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
- :31282-04-9
- :C₂₀H₃₇N₃O₁₃
- :527,52 g/mol
- Packung:Bottle