Biotin ≥98%
Bottle
Katalognr. APOSBUP18500-10G
New Product
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
Usage: Biochemical/ life science applications.
- Store at 2 to 8 °C Non-hazardous
Synonyme: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- CAS Nummer:58-85-5
- Formel:C₁₀H₁₆N₂O₃S
- Molekulargewicht:244,31 g/mol
- Lagertemperatur:Kühlschrank
- EINECS:200-399-3
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



