Biotin ≥98%
Bottle
Katalognr. APOSBUP18500-1G
New Product
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
Usage: Biochemical/ life science applications.
- Store at 2 to 8 °C Non-hazardous
Synonyme: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- CAS Nummer:58-85-5
- Formel:C₁₀H₁₆N₂O₃S
- Molekulargewicht:244,31 g/mol
- Lagertemperatur:Kühlschrank
- EINECS:200-399-3
Technische Daten
Technische Daten
- Packung:Bottle



