Shield-1 ≥98%
Bottle
Katalognr. APOSBUP09132-50MG
New Product
Anbieter: Apollo Scientific
Shield-1 ≥98%
Katalognr. APOSBUP09132-50MG
Anbieter: Apollo Scientific
New Product
CAS Nummer: 914805-33-7
Direkt zu
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyme: Shield-1
- CAS Nummer:914805-33-7
- Formel:C₄₂H₅₆N₂O₁₀
- Molekulargewicht:748,9 g/mol
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



