(-)-β-pinene ≥95%
Bottle
Catalog # APOSBUP19725-10G
New Product
Supplier: Apollo Scientific
(-)-β-pinene ≥95%
Catalog # APOSBUP19725-10G
Supplier: Apollo Scientific
New Product
CAS Number: 18172-67-3
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: (-)-Pin-2(10)-ene
- CAS Number:18172-67-3
- Formula:C₁₀H₁₆
- Molecular Weight:136,23 g/mol
Specifications
Specifications
- Packung:Bottle



