L-Ascorbic acid sodium salt
Bottle
Katalognr. APOSBUP18276-500MG
New Product
Anbieter: Apollo Scientific
L-Ascorbic acid sodium salt
Katalognr. APOSBUP18276-500MG
Anbieter: Apollo Scientific
New Product
CAS Nummer:
Direkt zu
Product Details & Documents
Synonyme: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
- Molekulargewicht:198,11 g/mol
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



