Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Colchicine ≥98%

Bottle
  APOSBUP16899-200MG
New Product
Colchicine ≥98%
  APOSBUP16899-200MG
New Product
 :  64-86-8
Colchicine ≥98%

Bottle

 :  64-86-8
 
SFr. 76.20
   
1 * 200 mg
 

Usage: Biochemical/ life science applications.

 :  7α-H-Colchicine, (S)-(-)-Colchicine
  •  :64-86-8
  •  :C₂₂H₂₅NO₆
  •  :399,44 g/mol
 

 

  • Packung:
    Bottle

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.