Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
DAPT ≥98%

DAPT ≥98%

Bottle
Catalog # APOSBUP15441-200MG
New Product
Supplier:   Apollo Scientific
DAPT ≥98%
Catalog # APOSBUP15441-200MG
Supplier:  Apollo Scientific
New Product
CAS Number:  208255-80-5
DAPT ≥98%

Bottle

CAS Number:  208255-80-5
Price
830.00 SFr.
Log in for availability
1 * 200 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, tert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate, (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester
  • CAS Number:208255-80-5
  • Formula:C₂₃H₂₆F₂N₂O₄
  • Molecular Weight:432,46 g/mol
Specifications

Specifications

  • Packung:
    Bottle

Specifications