Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Dihydroethidium ≥95%

Bottle
  APOSBUP15530-100MG
New Product
Dihydroethidium ≥95%
  APOSBUP15530-100MG
New Product
 :  104821-25-2
Dihydroethidium ≥95%

Bottle

 :  104821-25-2
 
279.00 SFr.
   
1 * 100 mg
 

Usage: Biochemical/life science applications.

 :  3,8-Diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine, Hydroethidine, DHE, 5-Ethyl-6-phenyl-5,6-dihydrophenanthridine-3,8-diamine, 2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine
  •  :104821-25-2
  •  :C₂₁H₂₁N₃
  •  :315,41 g/mol
 

 

  • Packung:
    Bottle

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.