Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Copper Pyrophosphate 98%

Bottle
  APOSIN11914-100G
New Product
Copper Pyrophosphate 98%
  APOSIN11914-100G
New Product
 :  10102-90-6
Copper Pyrophosphate 98%

Bottle

 :  10102-90-6
 
SFr. 29.50
   
1 * 100 g
 

Usage: Inorganic salt or catalyst

  •  :10102-90-6
  •  :Cu₂O₇P₂
  •  :301,04 g/mol
 

 

  • Packung:
    Bottle

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.