DEAE-Dextran, 70 kDa
Bottle
Katalognr. APOSOR1027098-5G
New Product
Anbieter: Apollo Scientific
DEAE-Dextran, 70 kDa
Katalognr. APOSOR1027098-5G
Anbieter: Apollo Scientific
New Product
CAS Nummer: 9015-73-0
Direkt zu
Product Details & Documents
Usage: Organic building block/reagent
Synonyme: Diethylaminoethyl dextran
- CAS Nummer:9015-73-0
Technische Daten
Technische Daten
- Packung:Bottle