Deoxythymidine triphosphate ≥95%
Bottle
Catalog # APOSBUP05081-200MG
New Product
Supplier: Apollo Scientific
Deoxythymidine triphosphate ≥95%
Catalog # APOSBUP05081-200MG
Supplier: Apollo Scientific
New Product
CAS Number: 18423-43-3
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: Thymidine-5'-triphosphoric acid, Deoxythymidine triphosphate, TPP, 2'-Deoxythymidine-5'-triphosphoric acid, dTTP
- CAS Number:18423-43-3
- Formula:C₁₀H₁₄N₂Na₃O₁₄P₃
- Molecular Weight:548,11 g/mol
Specifications
Specifications
- Packung:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



