Deoxythymidine triphosphate ≥95%
Bottle
Katalognr. APOSBUP05081-100MG
New Product
Anbieter: Apollo Scientific
Deoxythymidine triphosphate ≥95%
Katalognr. APOSBUP05081-100MG
Anbieter: Apollo Scientific
New Product
CAS Nummer: 18423-43-3
Direkt zu
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyme: Thymidine-5'-triphosphoric acid, Deoxythymidine triphosphate, TPP, 2'-Deoxythymidine-5'-triphosphoric acid, dTTP
- CAS Nummer:18423-43-3
- Formel:C₁₀H₁₄N₂Na₃O₁₄P₃
- Molekulargewicht:548,11 g/mol
Technische Daten
Technische Daten
- Packung:Bottle



