DAPT
Bottle
APOSBISN0140-250MG
New Product
DAPT is an inhibitor of -secretase and it can reduce the expression of A40 and A42 with IC50 values of 115 nM and 200 nM.
: N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, tert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate, (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester
- :208255-80-5
- :C₂₃H₂₆F₂N₂O₄
- :MFCD04974585
- :432.47 g/mol
- :Raumtemperatur
- Packung:Bottle
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.