AuftragsverfolgungKontakt
DAPT
Bottle
  APOSBISN0140-250MG
New Product
DAPT
  APOSBISN0140-250MG
New Product
 :  208255-80-5
DAPT
Bottle
 :  208255-80-5
SFr. 383.00
1 * 250 mg
DAPT is an inhibitor of -secretase and it can reduce the expression of A40 and A42 with IC50 values of 115 nM and 200 nM.
 :  N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, tert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate, (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester
  •  :208255-80-5
  •  :C₂₃H₂₆F₂N₂O₄
  •  :MFCD04974585
  •  :432.47 g/mol
  •  :Raumtemperatur
  • Packung:
    Bottle