Natriumpyruvat
Bottle
Katalognr. APOSBIB6270-25KG
New Product
Anbieter: Apollo Scientific
Natriumpyruvat
Katalognr. APOSBIB6270-25KG
Anbieter: Apollo Scientific
New Product
CAS Nummer: 113-24-6
Direkt zu
Product Details & Documents
Medium of animal cell culture; Material for pharmaceuticals, agrochemicals and diagnostic reagent; Intermediate for flavors and fragrances.
Synonyme: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS Nummer:113-24-6
- Formel:CH₃COCOONa
- MDL-Nummer:MFCD00002586
- Molekulargewicht:110.04 g/mol
- Schmelzender Platingehalt:>300 °C
- Schmelzender Platingehalt:>300 °C
- Lagertemperatur:Kühlschrank
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



