Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Natriumpyruvat

Natriumpyruvat

Bottle
Catalog # APOSBIB6270-25G
Irritant
New Product
Supplier:   Apollo Scientific
Natriumpyruvat
Catalog # APOSBIB6270-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  113-24-6
Natriumpyruvat

Bottle

CAS Number:  113-24-6
Price
SFr. 26.00
Log in for availability
1 * 25 g
Product Details & Documents

Medium of animal cell culture; Material for pharmaceuticals, agrochemicals and diagnostic reagent; Intermediate for flavors and fragrances.

Synonyms:  2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
  • CAS Number:113-24-6
  • Formula:CH₃COCOONa
  • MDL Number:MFCD00002586
  • Molecular Weight:110.04 g/mol
  • Melting Pt:>300 °C
  • Melting Pt:>300 °C
  • Storage Temperature:Kühlschrank
Specifications

Specifications

  • Packung:
    Bottle

Specifications